4-Methoxy-2-methylbenzaldehyde 98%

BLDPBD6666-5GEA 39.5 EUR
BLDPBD6666-5G BLDPBD6666-25G BLDPBD6666-100G BLDPBD6666-1G
4-Methoxy-2-methylbenzaldehyde 98%
4-Methoxy-2-methylbenzaldehyde

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₉H₁₀O₂
MW: 150,18 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD02261771
CAS-numero: 52289-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR