2,4-Dimethoxybenzenesulphonamide 96%

BLDPBD66996-25GEA 398 EUR
BLDPBD66996-25G BLDPBD66996-5G BLDPBD66996-1G
2,4-Dimethoxybenzenesulphonamide 96%
2,4-Dimethoxybenzenesulphonamide

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₈H₁₁NO₄S
MW: 217,25 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD06147001
CAS-numero: 51770-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR