Bis(isothiocyanato)(2,2'-bipyridyl-4,4'-dicarboxylato)(4,4'-dinonyl-2,2'-bipyridyl)ruthenium(II) 98%

BLDPBD682506-100MGEA 88.4 EUR
BLDPBD682506-100MG BLDPBD682506-5G BLDPBD682506-250MG BLDPBD682506-1G
Bis(isothiocyanato)(2,2'-bipyridyl-4,4'-dicarboxylato)(4,4'-dinonyl-2,2'-bipyridyl)ruthenium(II) 98%
Bis(isothiocyanato)(2,2'-bipyridyl-4,4'-dicarboxylato)(4,4'-dinonyl-2,2'-bipyridyl)ruthenium(II)

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₄₂H₅₂N₆O₄RuS₂
MW: 870,11 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD12546029
CAS-numero: 502693-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR