1-(2-Furoyl)piperazine hydrochloride 95%

BLDPBD6904-25GEA 115 EUR
BLDPBD6904-25G BLDPBD6904-100G BLDPBD6904-1G BLDPBD6904-5G BLDPBD6904-10G
1-(2-Furoyl)piperazine hydrochloride 95%
1-(2-Furoyl)piperazine hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₉H₁₃ClN₂O₂
MW: 216,6647 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD00236012
CAS-numero: 60548-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR