(S)-β-Phenylalaninol 98%

BLDPBD6964-100MGEA 6.3 EUR
BLDPBD6964-100MG BLDPBD6964-25G BLDPBD6964-5G BLDPBD6964-10G BLDPBD6964-1G BLDPBD6964-100G BLDPBD6964-250MG
(S)-β-Phenylalaninol 98%
(S)-β-Phenylalaninol

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Kaava: C₉H₁₃NO
MW: 151,2056 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD01311768
CAS-numero: 82769-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR