Ethyl-2-amino-4-phenylthiophene-3-carboxylate 96%

BLDPBD70270-5GEA 71.7 EUR
BLDPBD70270-5G BLDPBD70270-250MG BLDPBD70270-1G BLDPBD70270-25G
Ethyl-2-amino-4-phenylthiophene-3-carboxylate 96%
Ethyl-2-amino-4-phenylthiophene-3-carboxylate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Kaava: C₁₃H₁₃NO₂S
MW: 247,32 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD00126391
CAS-numero: 4815-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR