2-Bromo-N,N-diethylbenzamide 97%

BLDPBD72234-25GEA 192 EUR
BLDPBD72234-25G BLDPBD72234-5G BLDPBD72234-1G
2-Bromo-N,N-diethylbenzamide 97%
2-Bromo-N,N-diethylbenzamide

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₁H₁₄BrNO
MW: 256,1390 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00457139
CAS-numero: 76041-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR