2-Bromo-N-methylbenzenesulphonamide 98%

BLDPBD72252-1GEA 83.2 EUR
BLDPBD72252-1G BLDPBD72252-250MG
2-Bromo-N-methylbenzenesulphonamide 98%
2-Bromo-N-methylbenzenesulphonamide

Storage condition: Sealed in dry, Room Temperature

Kaava: C₇H₈BrNO₂S
MW: 250,11 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD03084756
CAS-numero: 98192-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR