2-Chloro-4-(fur-2-yl)pyrimidine 97%

BLDPBD72830-1GEA 220 EUR
BLDPBD72830-1G BLDPBD72830-100MG BLDPBD72830-250MG
2-Chloro-4-(fur-2-yl)pyrimidine 97%
2-Chloro-4-(fur-2-yl)pyrimidine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Kaava: C₈H₅ClN₂O
MW: 180,591 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD09065030
CAS-numero: 124959-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR