2-Chloro-3'-methoxyacetanilide 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD73751-5GEA 259 EUR
BLDPBD73751-5G BLDPBD73751-1G BLDPBD73751-250MG
2-Chloro-3'-methoxyacetanilide 98%
2-Chloro-3'-methoxyacetanilide

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₉H₁₀ClNO₂
MW: 199,6342 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00018900
CAS-numero: 17641-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR