Ethyl-2-ethylbutyrate 98%

BLDPBD74710-1GEA 7.3 EUR
BLDPBD74710-1G BLDPBD74710-5G
Ethyl-2-ethylbutyrate 98%
Ethyl-2-ethylbutyrate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₈H₁₆O₂
MW: 144,21 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00059396
CAS-numero: 2983-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR