3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate 97%
|
|
Synonyms:
Dihydrochloride TMB
, TMB-HCl
BLDPBD748724-10GEA
59.3
EUR
BLDPBD748724-10G
BLDPBD748724-1G
BLDPBD748724-250MG
BLDPBD748724-25G
BLDPBD748724-5G
3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate 97%
3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Kaava:
C₁₆H₂₀N₂·2HCl·xH₂O MW: 313,27 g/mol (anhy) Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD00150104 CAS-numero: 207738-08-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...