1,3,6,8-Tetramethyl-9H-carbazole 98%
|
|
BLDPBD749037-100MGEA
52
EUR
BLDPBD749037-100MG
1,3,6,8-Tetramethyl-9H-carbazole 98%
1,3,6,8-Tetramethyl-9H-carbazole
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Kaava:
C₁₆H₁₇N MW: 223,32 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD18450169 CAS-numero: 6558-85-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...