4-Isoquinolineboronic acid 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD7504-5GEA 149 EUR
BLDPBD7504-5G BLDPBD7504-1G BLDPBD7504-25G
4-Isoquinolineboronic acid 98%
4-Isoquinolineboronic acid

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Kaava: C₉H₈BNO₂
MW: 172,9763 g/mol
Storage Temperature: Pakastin
MDL numero: MFCD03412093
CAS-numero: 192182-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR