2-Azaspiro[4.5]decane hydrochloride 98%

BLDPBD758187-100MGEA 152 EUR
BLDPBD758187-100MG
2-Azaspiro[4.5]decane hydrochloride 98%
2-Azaspiro[4.5]decane hydrochloride

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Kaava: C₉H₁₇N·ClH
MW: 175,699 g/mol
Storage Temperature: Pakastin
MDL numero: MFCD05861558
CAS-numero: 36392-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR