2-Isobutyrylcyclohexan-1-one 97%

BLDPBD76092-5GEA 113 EUR
BLDPBD76092-5G BLDPBD76092-25G BLDPBD76092-10G BLDPBD76092-0.025G BLDPBD76092-100G BLDPBD76092-1G
2-Isobutyrylcyclohexan-1-one 97%
2-Isobutyrylcyclohexan-1-one

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₀H₁₆O₂
MW: 168,233 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00466185
CAS-numero: 39207-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR