2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98% (by GC)
|
|
BLDPBD76350-25GEA
19.8
EUR
BLDPBD76350-25G
BLDPBD76350-5G
BLDPBD76350-10G
BLDPBD76350-1G
2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98% (by GC)
2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Storage condition: Inert atmosphere, 2-8°C
Kaava:
C₇H₁₅BO₃ MW: 158,01 g/mol Storage Temperature: Jääkaappi |
MDL numero:
MFCD03093086 CAS-numero: 1195-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...