(4-Bromophenoxy)trimethylsilane 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD763969-100GEA 273 EUR
BLDPBD763969-100G BLDPBD763969-5G BLDPBD763969-1G BLDPBD763969-25G
(4-Bromophenoxy)trimethylsilane 98%
(4-Bromophenoxy)trimethylsilane

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₉H₁₃BrOSi
MW: 245,20 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00040713
CAS-numero: 17878-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR