2-Methyl-1,3-thiazole-4-carbonitrile 97%

BLDPBD76640-5GEA 375 EUR
BLDPBD76640-5G BLDPBD76640-250MG BLDPBD76640-1G BLDPBD76640-100MG
2-Methyl-1,3-thiazole-4-carbonitrile 97%
2-Methyl-1,3-thiazole-4-carbonitrile

Storage condition: Sealed in dry, 2-8°C

Kaava: C₅H₄N₂S
MW: 124,164 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD00068103
CAS-numero: 21917-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR