2-Methyl-4(1H)-quinazolinone 95%

BLDPBD76911-10GEA 113 EUR
BLDPBD76911-10G BLDPBD76911-1G BLDPBD76911-25G BLDPBD76911-5G BLDPBD76911-250MG
2-Methyl-4(1H)-quinazolinone 95%
2-Methyl-4(1H)-quinazolinone

Storage condition: Sealed in dry, Room Temperature

Kaava: C₉H₈N₂O
MW: 160,18 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD05270928
CAS-numero: 1769-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR