2-Methyl-5-iodopyridine 97%

BLDPBD77035-250MGEA 56.2 EUR
BLDPBD77035-250MG BLDPBD77035-1G BLDPBD77035-25G BLDPBD77035-5G BLDPBD77035-10G BLDPBD77035-0.025G
2-Methyl-5-iodopyridine 97%
2-Methyl-5-iodopyridine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Kaava: C₆H₆IN
MW: 219,023 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD07437990
CAS-numero: 695-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR