5'-Fluoro-2'-methoxyacetophenone 98%
|
|
BLDPBD7737-25GEA
173
EUR
BLDPBD7737-25G
BLDPBD7737-1G
BLDPBD7737-10G
BLDPBD7737-5G
5'-Fluoro-2'-methoxyacetophenone 98%
5'-Fluoro-2'-methoxyacetophenone
Storage condition: Sealed in dry, Room Temperature
Kaava:
C₉H₉FO₂ MW: 168,17 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD00671763 CAS-numero: 445-82-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...