Methyl-3-methyl-4-nitrobenzoate 98%

BLDPBD7824-100GEA 61.3 EUR
BLDPBD7824-100G BLDPBD7824-25G BLDPBD7824-500G BLDPBD7824-10G BLDPBD7824-5G
Methyl-3-methyl-4-nitrobenzoate 98%
Methyl-3-methyl-4-nitrobenzoate

Storage condition: Sealed in dry, Room Temperature

Kaava: C₉H₉NO₄
MW: 195,18 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00085640
CAS-numero: 24078-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR