2-(2-Hydroxyprop-2-yl)pyridine 97%
|
|
BLDPBD78456-250MGEA
104
EUR
BLDPBD78456-250MG
BLDPBD78456-100MG
2-(2-Hydroxyprop-2-yl)pyridine 97%
2-(2-Hydroxyprop-2-yl)pyridine
Storage condition: Inert atmosphere, Room Temperature
Kaava:
C₈H₁₁NO MW: 137,179 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD00209259 CAS-numero: 37988-38-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...