3-(4-Chlorophenoxy)propanenitrile 95%

BLDPBD79990-250MGEA 28.1 EUR
BLDPBD79990-250MG BLDPBD79990-5G BLDPBD79990-1G
3-(4-Chlorophenoxy)propanenitrile 95%
3-(4-Chlorophenoxy)propanenitrile

Storage condition: Sealed in dry, Room Temperature

Kaava: C₉H₈ClNO
MW: 181,619 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01569392
CAS-numero: 46125-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR