5-Amino-3-(4-chlorophenyl)isoxazole 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD80025-100MGEA 41.6 EUR
BLDPBD80025-100MG BLDPBD80025-1G BLDPBD80025-5G BLDPBD80025-250MG
5-Amino-3-(4-chlorophenyl)isoxazole 98%
5-Amino-3-(4-chlorophenyl)isoxazole

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₉H₇ClN₂O
MW: 194,6177 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01569450
CAS-numero: 33866-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR