2-Chloro-3-(trifluoromethyl)phenylboronic acid 97%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD8018-1GEA 17.7 EUR
BLDPBD8018-1G BLDPBD8018-25G BLDPBD8018-5G BLDPBD8018-250MG BLDPBD8018-10G
2-Chloro-3-(trifluoromethyl)phenylboronic acid 97%
2-Chloro-3-(trifluoromethyl)phenylboronic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₇H₅BClF₃O₂
MW: 224,3726 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD09258745
CAS-numero: 957061-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR