3,5-Dibromopyridine-N-oxide 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD82458-5GEA 65.5 EUR
BLDPBD82458-5G BLDPBD82458-25G BLDPBD82458-1G
3,5-Dibromopyridine-N-oxide 98%
3,5-Dibromopyridine-N-oxide

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₅H₃Br₂NO
MW: 252,8914 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00160750
CAS-numero: 2402-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR