4-Carboxy-3-chlorophenylboronic acid 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD8271-100MGEA 6.3 EUR
BLDPBD8271-100MG BLDPBD8271-250MG BLDPBD8271-1G
4-Carboxy-3-chlorophenylboronic acid 98%
4-Carboxy-3-chlorophenylboronic acid

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₇H₆BClO₄
MW: 200,39 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD06656268
CAS-numero: 136496-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR