3',5'-Dimethyl-2'-hydroxyacetophenone 98%

BLDPBD82796-10GEA 114 EUR
BLDPBD82796-10G BLDPBD82796-5G BLDPBD82796-25G BLDPBD82796-1G BLDPBD82796-100G
3',5'-Dimethyl-2'-hydroxyacetophenone 98%
3',5'-Dimethyl-2'-hydroxyacetophenone

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₁₀H₁₂O₂
MW: 164,2000 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01631133
CAS-numero: 1198-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR