3-Amino-3-(2-methylphenyl)propionic acid 98%
|
|
BLDPBD83887-25GEA
1320
EUR
BLDPBD83887-25G
BLDPBD83887-1G
BLDPBD83887-250MG
BLDPBD83887-5G
3-Amino-3-(2-methylphenyl)propionic acid 98%
3-Amino-3-(2-methylphenyl)propionic acid
Storage condition: Keep in dark place, Sealed in dry, 2-8°C
Kaava:
C₁₀H₁₃NO₂ MW: 179,2200 g/mol Storage Temperature: Jääkaappi |
MDL numero:
MFCD01871311 CAS-numero: 68208-16-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...