5-Hydroksi-isoftaalihappo 98%
|
|
BLDPBD8414-500GEA
176
EUR
BLDPBD8414-500G
BLDPBD8414-25G
BLDPBD8414-100G
BLDPBD8414-1000G
BLDPBD8414-10G
5-Hydroksi-isoftaalihappo 98%
5-Hydroksi-isoftaalihappo
Storage condition: Inert atmosphere, Room Temperature
Kaava:
C₈H₆O₅ MW: 182,13 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD00002515 CAS-numero: 618-83-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...