3-Bromo-5-ethoxy-4-methoxybenzonitrile 97%

BLDPBD85179-5GEA 493 EUR
BLDPBD85179-5G BLDPBD85179-1G BLDPBD85179-250MG
3-Bromo-5-ethoxy-4-methoxybenzonitrile 97%
3-Bromo-5-ethoxy-4-methoxybenzonitrile

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₀H₁₀BrNO₂
MW: 256,0959 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD02256583
CAS-numero: 515831-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR