N-Boc-L-tyrosine 98%

BLDPBD8539-10GEA 9.4 EUR
BLDPBD8539-10G BLDPBD8539-100G BLDPBD8539-25G BLDPBD8539-5G BLDPBD8539-500G
N-Boc-L-tyrosine 98%
N-Boc-L-tyrosine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₁₄H₁₉NO₅
MW: 281,3 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00037179
CAS-numero: 3978-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR