3-Chloropropanamide 98%

BLDPBD86185-5GEA 73.8 EUR
BLDPBD86185-5G BLDPBD86185-25G BLDPBD86185-1G
3-Chloropropanamide 98%
3-Chloropropanamide

Storage condition: Sealed in dry, Room Temperature

Kaava: C₃H₆ClNO
MW: 107,539 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008040
CAS-numero: 5875-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR