3-Ethoxy-4-hydroxy-5-nitrobenzaldehyde 95%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD86526-1GEA 42.6 EUR
BLDPBD86526-1G BLDPBD86526-5G BLDPBD86526-250MG
3-Ethoxy-4-hydroxy-5-nitrobenzaldehyde 95%
3-Ethoxy-4-hydroxy-5-nitrobenzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₉H₉NO₅
MW: 211,1715 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD00058678
CAS-numero: 178686-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR