Methyl-4-methoxysalicylate 98%

BLDPBD8682-500GEA 450 EUR
BLDPBD8682-500G BLDPBD8682-5G BLDPBD8682-10G BLDPBD8682-0.5G BLDPBD8682-25G BLDPBD8682-100G
Methyl-4-methoxysalicylate 98%
Methyl-4-methoxysalicylate

Storage condition: Sealed in dry, Room Temperature

Kaava: C₉H₁₀O₄
MW: 182,18 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008424
CAS-numero: 5446-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR