3-Iodo-6-nitro-1H-indazole 97%
|
|
BLDPBD87496-1GEA
26
EUR
BLDPBD87496-1G
BLDPBD87496-25G
BLDPBD87496-250MG
BLDPBD87496-5G
BLDPBD87496-0.025G
3-Iodo-6-nitro-1H-indazole 97%
3-Iodo-6-nitro-1H-indazole
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Kaava:
C₇H₄IN₃O₂ MW: 289,0300 g/mol Storage Temperature: Jääkaappi |
MDL numero:
MFCD07781652 CAS-numero: 70315-70-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...