3-Nitrobiphenyl 98%

BLDPBD88548-5GEA 55.1 EUR
BLDPBD88548-5G BLDPBD88548-250MG BLDPBD88548-25G BLDPBD88548-1G
3-Nitrobiphenyl 98%
3-Nitrobiphenyl

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₂H₉NO₂
MW: 199,21 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00007245
CAS-numero: 2113-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR