4-(2-Bromoethyl)tetrahydropyran 97%
|
|
BLDPBD89819-250MGEA
73.8
EUR
BLDPBD89819-250MG
BLDPBD89819-5G
BLDPBD89819-100MG
BLDPBD89819-1G
4-(2-Bromoethyl)tetrahydropyran 97%
4-(2-Bromoethyl)tetrahydropyran
Storage condition: Sealed in dry, Room Temperature
Kaava:
C₇H₁₃BrO MW: 193,0800 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD09800414 CAS-numero: 4677-20-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...