4,5-Difluoro-o-toluidine 95%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD92562-25GEA 217 EUR
BLDPBD92562-25G BLDPBD92562-1G BLDPBD92562-10G BLDPBD92562-5G BLDPBD92562-250MG
4,5-Difluoro-o-toluidine 95%
4,5-Difluoro-o-toluidine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Kaava: C₇H₇F₂N
MW: 143,14 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD08062378
CAS-numero: 875664-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR