4'-Acetamidoacetophenone 98%
|
|
BLDPBD93344-1GEA
6.3
EUR
BLDPBD93344-1G
BLDPBD93344-5G
BLDPBD93344-100G
BLDPBD93344-25G
4'-Acetamidoacetophenone 98%
4'-Acetamidoacetophenone
Storage condition: Inert atmosphere, Room Temperature
Kaava:
C₁₀H₁₁NO₂ MW: 177,20 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD00014965 CAS-numero: 2719-21-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...