4-Bromo-4'-tert-butylbiphenyl 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD95110-25GEA 161 EUR
BLDPBD95110-25G BLDPBD95110-1G BLDPBD95110-5G
4-Bromo-4'-tert-butylbiphenyl 98%
4-Bromo-4'-tert-butylbiphenyl

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₆H₁₇Br
MW: 289,21 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01321141
CAS-numero: 162258-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR