4-Bromostilbene 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD95374-250MGEA 10.4 EUR
BLDPBD95374-250MG BLDPBD95374-25G BLDPBD95374-1G BLDPBD95374-5G
4-Bromostilbene 98%
4-Bromostilbene

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₄H₁₁Br
MW: 259,1411 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00155020
CAS-numero: 4714-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR