Methyl-2-aminothiazole-4-carboxylate 97%
|
|
BLDPBD9603-500GEA
336
EUR
BLDPBD9603-500G
BLDPBD9603-5G
BLDPBD9603-25G
BLDPBD9603-100G
BLDPBD9603-1G
BLDPBD9603-10G
Methyl-2-aminothiazole-4-carboxylate 97%
Methyl-2-aminothiazole-4-carboxylate
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Kaava:
C₅H₆N₂O₂S MW: 158,1800 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD00622441 CAS-numero: 118452-04-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...