(S)-(+)-2,2-Dimethylcyclopropanecarboxamide 98%
|
|
BLDPBD9656-5GEA
7.3
EUR
BLDPBD9656-5G
BLDPBD9656-1G
BLDPBD9656-25G
BLDPBD9656-500G
BLDPBD9656-10G
BLDPBD9656-100G
(S)-(+)-2,2-Dimethylcyclopropanecarboxamide 98%
(S)-(+)-2,2-Dimethylcyclopropanecarboxamide
Storage condition: Sealed in dry, Room Temperature
Kaava:
C₆H₁₁NO MW: 113,1600 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD00216614 CAS-numero: 75885-58-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...