2-Bromo-4'-fluoro-3'-(trifluoromethyl)acetophenone 97%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD97175-1GEA 13.6 EUR
BLDPBD97175-1G BLDPBD97175-10G BLDPBD97175-5G BLDPBD97175-100G BLDPBD97175-25G
2-Bromo-4'-fluoro-3'-(trifluoromethyl)acetophenone 97%
2-Bromo-4'-fluoro-3'-(trifluoromethyl)acetophenone

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₉H₅BrF₄O
MW: 285,0330 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD03094509
CAS-numero: 537050-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR