4-Fluoro-3-nitrobenzenesulphonyl chloride 95%

BLDPBD97220-25GEA 568 EUR
BLDPBD97220-25G BLDPBD97220-1G BLDPBD97220-250MG BLDPBD97220-5G
4-Fluoro-3-nitrobenzenesulphonyl chloride 95%
4-Fluoro-3-nitrobenzenesulphonyl chloride

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₆H₃ClFNO₄S
MW: 239,6100 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD06342770
CAS-numero: 6668-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR