4-Hydroxy-3-methoxybenzylamine hydrochloride 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD97692-100GEA 71.7 EUR
BLDPBD97692-100G BLDPBD97692-5G BLDPBD97692-25G BLDPBD97692-10G
4-Hydroxy-3-methoxybenzylamine hydrochloride 98%
4-Hydroxy-3-methoxybenzylamine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₈H₁₂ClNO₂
MW: 189,64 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00012864
CAS-numero: 7149-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR