Ethyl (4-iodophenyl)acetate 95%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD98158-10GEA 123 EUR
BLDPBD98158-10G BLDPBD98158-5G BLDPBD98158-25G BLDPBD98158-100G BLDPBD98158-250MG BLDPBD98158-1G
Ethyl (4-iodophenyl)acetate 95%
Ethyl (4-iodophenyl)acetate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Kaava: C₁₀H₁₁IO₂
MW: 290,0976 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD07779004
CAS-numero: 15250-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR